Learn More
Cytiva HyClone™ HyCell-STEM™ Media
Complete media for culturing human embryonic stem cells (hESCs) and induced pluripotent stem (hiPS) cells.
Brand: Cytiva SR30003.01
Code : NEW
Additional Details : Weight : 1.00000kg
Alert:
Hyclone Cell Culture products now a part of GE Healthcare.
Includes: HyCell-STEM kit includes 6x supplement and DMEM/F12 to create a complete media. Basic fibroblast growth factor must be added to the completed media at time of use.
Requires : bFGF (10ng/ml) must be added to the media at time of use.
Description
HyCell-STEM Media is designed to help generate large numbers of human embryonic and induced pluripotent stem cells with fewer passages and is intended for use with stem cells on feeder layer cultures. HyCell-STEM is provided as a 6x supplement that is added to DMEM/F12 to create a complete media. Basic fibroblast growth factor is added to the completed media at time of use.
Highlights:
- Increase rate of expansion
- Sustain healthy stem cell morphology
- Maintain stemness
- Preserve pluripotency
- Improved post-thaw recovery
HyCell-STEM Media is intended for use with stem cells on feeder layer cultures.
Specifications
Stem Cell Supplement Media | |
Stem Cell Culture | |
100 mL |