Learn More
Cytiva HyClone™ HyCell TransFx-C Powdered Media
Animal-derived component-free (ADCF), hydrolysate-free, and regulatory-friendly cell culture medium
Brand: Cytiva SH30942.01
Additional Details : CAS Number : 7048-04-6 Weight : 0.14160kg
Description
The versatility of this medium allows quick adaptation and supports exceptional transfection, high viable cell density, and high productivity across a variety of Chinese hamster ovary (CHO) cell lines.
HyCell TransFx-C is available in liquid and powder formats in user-friendly packaging.
- ADCF and hydrolysate-free formulation
- Supports high transfection efficiency
- Designed for high cell yield and recombinant protein production
- Allows for direct or sequential adaptation
- Designed for micro- to large-scale transfection and production applications
- Manufactured from traceable components according to cGMP guidelines
Specifications
2 to 8°C, away from light and moisture | |
Non-sterile | |
5 L |
Powder | |
No Glutamine, No Poloxamer 188 |