Learn More
Cytiva HyClone™ HyCell TransFx-H Powdered Media
Animal-derived component-free (ADCF), hydrolysate-free, and regulatory-friendly cell culture medium.
Brand: Cytiva SH30944.01
282.15 GBP valid until 2024-12-31
Use promo code "23281" to get your promotional price.
Additional Details : CAS Number : 56-45-1 Weight : 0.11300kg
Description
The versatility of this medium allows quick adaptation and supports exceptional transfection, high viable cell density, and high productivity across a variety of human embryonic kidney (HEK) 293 cell lines.
HyCell TransFx-H is available in liquid and powder formats in user friendly packaging.
- Hydrolysate-free and ADCF formulation
- Supports high transfection efficiency
- Designed for high cell yield and recombinant protein production
- Allows for direct or sequential adaptation
- Designed for micro- to large-scale transfection and production applications
- Manufactured from traceable components according to cGMP guidelines
Specifications
2 to 8°C, away from light and moisture | |
Non-sterile | |
5 L |
Powder | |
No Glutamine, No Poloxamer 188 |