Learn More
Cytiva HyClone™ IMDM Media, Powder
Brand:  Cytiva SH30005.02
Product Code. 11918075
  • £88.80 / Pack of 2
Explore more special offers
Specifications
cGMP and ISO 9001:2000
IMDM with L-Glutamine, HEPES, without Alpha-Thioglycerol
IMDM with L-glutamine, HEPES; without alpha-thioglycerol
Glutamine, HEPES
2 x 5 L
2° to 8°C, away from light and moisture
Powder
Non-sterile
No α-Thioglycerol