missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Iron-Supplemented Bovine Calf Serum, US Origin
Contains additional iron that boosts serum transferrin levels to physiological levels
Brand: Cytiva SH30072.03IH
Code : 85
Additional Details : Weight : 0.50000kg
Description
- Supplemented with iron and growth-promoting factors
- Age of animal at time of collection approximately 16-24 weeks
- Virus panel testing according to 9 CFR 113.53
- Low in antibodies and high in growth factors
- Sourced in the USA with complete traceability back to original source
- A cost-effective alternative to FBS
Specifications
Manufactured in cGMP (21 CFR 820) Compliant and ISO9001 Certified Facilities | |
United States | |
500 mL | |
Formula-Fed Veal Calves | |
Virus Panel Testing According to 9 CFR 113.53 | |
Supplementing Cell Culture Growth | |
<10 mg/dL | |
HyClone™ |
16 to 22 weeks | |
Calf Bovine Serum | |
Sequential-filtered, 100 nm | |
Irradiated, Heat Inactivated | |
<10 EU/mL | |
Yes | |
Yes |