Learn More
Cytiva HyClone™ Iron-Supplemented Calf Serum, U.S. origin
A cost-effective alternative to Fetal Bovine Serum (FBS). Contains additional iron to boost serum transferrin to physiological levels.
Brand: Cytiva SH30072.04IR
Code : 85
Additional Details : Weight : 1.00000kg
Description
HyClone Iron-Supplemented Bovine Calf Serum, US Origin is collected by venipuncture from formula-fed veal calves, which produce exceptionally high levels of transferrin. This serum is carefully supplemented with iron to load serum transferrin levels to physiological levels. After supplementation, the serum contains three to four times as much available iron and transferrin as fetal bovine serum (FBS) or equine serum.
Specifications
Insulin, Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
1000 mL | |
Sequential-filtered, 100 nm | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus |
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
Calf Bovine Serum | |
Sterile | |
Gamma Irradiated | |
Yes |