missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Iron-Supplemented Newborn Bovine Calf Serum (New Zealand)
A cost-effective alternative to Fetal Bovine Serum (FBS).
£65.48 - £150.96
Specifications
Additional Assays | Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG |
---|---|
Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. |
Country of Origin | New Zealand |
Sterility | Sterile |
Sterilization Method | Sequential-filtered, 100 nm |
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | |||||
10761274
|
Cytiva
SH30626.02 |
500 mL | None | N/A | ||||||
12330043
|
Cytiva
SH30626.02IR |
500 mL | Irradiated | N/A | ||||||
11582830
|
Cytiva
SH30626.03 |
1000 mL | None | N/A | ||||||
12694217
|
Cytiva
SH30626.03HI |
1000 mL | Heat Inactivated | N/A | ||||||
12684217
|
Cytiva
SH30626.03IR |
1000 mL | Irradiated | N/A | ||||||
17236946
|
Cytiva
SH30626.03IR25-40 |
1000 mL | Irradiated (25 to 40 doses) | N/A | ||||||
Description
Specifications
Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG | |
New Zealand | |
Sequential-filtered, 100 nm | |
≤ 25 mg/dL | |
Yes |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
Sterile | |
≤ 25 EU/mL | |
HyClone™ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva HyClone™ Iron-Supplemented Newborn Bovine Calf Serum (New Zealand)