missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ New Zealand Newborn Bovine Serum
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
Description
A high quality alternative to FBS. The same collection and processing procedures, including venipuncture are used in production of this serum.
Specifications
Specifications
| Additional Assays | Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG |
| Certifications/Compliance | USDA Regulations |
| Content And Storage | <–1°C |
| Product Type | Newborn Calf Bovine Serum |
| Quantity | 1000 mL |
| Sterility | Sterile |
| Sterilization Method | 0.1 μm filtration |
| Processing | Heat Inactivated |
| Tested For | Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus |
| Endotoxin Concentration | ≤25EU/mL |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion