missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Newborn Bovine Calf Serum
Supplement cell culture growth with New Zealand sourced Cytiva HyClone Newborn Bovine Calf Serum, an excellent and cost-effective alternative to fetal bovine serum (FBS).
£173.00
Specifications
Content And Storage | -10°C or Lower, Once Thawed Store at 2°C to 8°C |
---|---|
Country of Origin | New Zealand |
Product Type | Newborn Calf Bovine Serum |
Volume (Metric) | 1000 mL |
For Use With (Application) | Enhance Growth of Cells |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17682075
|
Cytiva
SH30401.02IH25-40 |
25 to 40 Doses |
£173.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17692075
|
Cytiva
SH30401.02IH30-45 |
30 to 45 Doses |
£173.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Supplement cell culture growth with New Zealand sourced Cytiva HyClone Newborn Bovine Calf Serum, an excellent and cost-effective alternative to fetal bovine serum (FBS).
- Sourced from animals that are typically less than 10 days old
- Traceable
- Conforms to USDA regulations
Specifications
-10°C or Lower, Once Thawed Store at 2°C to 8°C | |
Newborn Calf Bovine Serum | |
Enhance Growth of Cells |
New Zealand | |
1000 mL |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva HyClone™ Newborn Bovine Calf Serum