missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Newborn Bovine Calf Serum, Heat Inactivated
Augment cell culture growth with this cost-effective alternative to Fetal Bovine Serum (FBS)
£42.60 - £238.00
Specifications
Content And Storage | Store at or below -10°C |
---|---|
Age | <10 days |
Country of Origin | United States |
Form | Liquid |
Product Type | Newborn Calf Bovine Serum |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12694237
|
Cytiva
SH30118.02HI |
100 mL |
£42.60
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12664237
|
Cytiva
SH30118.03HI |
500 mL |
£134.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12654207
|
Cytiva
SH30118.04HI |
1000 mL |
£238.00
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Produced using careful collection and processing procedures, including venipuncture, to provide the highest quality product available
- Ensures excellent culture results without fear of contamination
- HyClone™ maintains complete control of the entire collection and production process for 100% traceability
- Newborn bovine calf serum undergoes the same careful collection and processing procedures (including venipuncture) used for other bovine serum products
- Sourced from the United States with complete traceability back to original source
- Age of animal at time of collection less than 10 days old
- Low in antibodies and high in growth factors
- Virus panel testing according to 9 CFR 113.53
Specifications
Store at or below -10°C | |
United States | |
Newborn Calf Bovine Serum | |
Sequential-filtered, 100 nm | |
≤ 50 EU/mL | |
Yes | |
HyClone™ |
<10 days | |
Liquid | |
Sterile | |
Heat Inactivated | |
Augmenting Cell Culture Growth | |
≤ 25 mg/dL | |
Frozen |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva HyClone™ Newborn Bovine Calf Serum, Heat Inactivated