missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Newborn Bovine Calf Serum, New Zealand Origin
An excellent and cost-effective alternative to Fetal Bovine Serum (FBS)
£86.75 - £151.00
Specifications
Content And Storage | Store in Freezer, <20°C |
---|---|
Age | <10 days |
Country of Origin | New Zealand |
Form | Serum |
Product Type | Newborn Calf Bovine Serum |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16211112
|
Cytiva
SH30401.01IR30-40 |
500 mL |
£86.75
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16241112
|
Cytiva
SH30401.02IR30-40 |
1000 mL |
£151.00
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Supplements cell culture growth
- Sourced from animals that are typically less than 10 days old
- Manufactured in accordance with cGMP guidelines at an ISO 9001: 2008 accredited facility
- Conforms to USDA regulations
- Traceable
- Filtered
Specifications
Store in Freezer, <20°C | |
New Zealand | |
Newborn Calf Bovine Serum | |
Irradiated (30-40 kGy) | |
HyClone™ |
<10 days | |
Serum | |
Sequential-filtered, 100 nm | |
Supplementing Cell Culture Growth |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva HyClone™ Newborn Bovine Calf Serum, New Zealand Origin