missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Newborn Bovine Calf Serum, New Zealand Origin
A cost-effective alternative to Fetal Bovine Serum (FBS).
£71.00 - £151.00
Specifications
Certifications/Compliance | USDA Regulations |
---|---|
Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C |
Sterility | Sterile |
Endotoxin Concentration | ≤ 25 EU/mL |
Mycoplasma Detected | Yes |
Product Code | Brand | Processing | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Processing | Quantity | Price | Quantity & Availability | |||||
16201112
|
Cytiva
SH30401.01IR25-40 |
Irradiated, 25 to 40 doses | 500 mL |
£86.75
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
||||||||||
12624237
|
Cytiva
SH30401.02HI |
Heat inactivated | 1000 mL |
£151.00
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16221112
|
Cytiva
SH30401.01IR30-45 |
Irradiated, 30 to 45 doses | 500 mL |
£86.75
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16231112
|
Cytiva
SH30401.02IR25-40 |
Irradiated, 25 to 40 doses | 1000 mL |
£151.00
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12802956
|
Cytiva
SH30401.01IR |
Irradiated | 500 mL | N/A | ||||||
12339962
|
Cytiva
SH30401.01HI |
Heat Inactivated | 500 mL |
£85.35
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16251112
|
Cytiva
SH30401.02IR30-45 |
Irradiated, 30 to 45 doses | 1000 mL |
£151.00
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
USDA Regulations | |
Sterile | |
Yes |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C | |
≤ 25 EU/mL | |
Yes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva HyClone™ Newborn Bovine Calf Serum, New Zealand Origin