Learn More
Cytiva HyClone™ Non Essential Amino Acids (NEAA) 100X Solution
Cell culture is ideal for supplementation of serum- and protein-free media. Cytiva HyClone™ Non-Essential Amino Acids (NEAA) 100X supports the growth of cells that have high energy demands and synthesize large amounts of proteins and nucleic acids.
Brand: Cytiva SH30238.01
14.80 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 83
Additional Details : CAS Number : 7647-01-0 Weight : 0.13500kg
Description
Components:- L-Alanine: 890mg/L
- L-Asparagine-H20: 1500mg/L
- L-Aspartic acid: 1330mg/L
- L-Glutamic acid: 1470mg/L
- Glycine: 750mg/L
- L-Proline: 1150mg/L
- L-Serine: 1050mg/L
Specifications
cGMP | |
NEAA Solution, 100X | |
Liquid | |
Supplementation of serum- and protein-free media |
2–8°C | |
100 X | |
Sterile | |
100 mL |
Safety and Handling
- MEM Non-Essential Amino Acids Solution
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305-IF IN EYES:Immediately call a POISON CENTER/doctor..
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : hydrochloric acid