missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ PBS 1X Phosphate 0.0067M without Ca or Mg
HyClone™ PBS 1X Phosphate 0.0067M without Ca or Mg is a balanced salt solution used for multiple cell culture applications.
Brand: Cytiva SH30256.14
This item is not returnable.
View return policy
Description
- HyClone phosphate buffered saline (PBS) cell culture solution includes sodium chloride, sodium phosphate, and potassium without calcium or magnesium.
- No sodium pyruvate is added.
- HyClone sodium phosphate buffer is a balanced solution available as 1X and 10X PBS buffer with 0.0067 M PO4, pH 7.0 to 7.2.
- Sterile buffered saline solution:
- HyClone phosphate buffered saline solutions are filtered through final 0.1 μm pore-size filters.
- Filter integrity tests are performed both before and after filtration.
- Aseptic fills are performed in a class 100 clean room.
- Each lot is subjected to sterility testing, which is performed using the membrane filtration method described in the current U.S. Pharmacopeia (USP).
Specifications
Phosphate Buffered Saline (PBS) | |
0.0067 M | |
Phosphate Buffered Saline (PBS) | |
0.1 μm Sterile-filtered | |
7.0 to 7.2 |
Clear, Colorless | |
Liquid | |
Sterile | |
Cell Culture | |
100 L |