Learn More
Cytiva™ HyClone™ Peak Expression Medium
Ready-to-use medium supports growth and efficient transfection of adeno-associated virus (AAV) expression in HEK 293 cell lines.
Brand: Cytiva™ SH31193.02
91.72 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 1.06400kg
Description
This medium supports high cell growth, high transfection efficiency, and robust viral production for a variety of AAV serotypes expressed in human embryonic kidney (HEK) cell lines. HyClone™ peak expression medium is an animal-derived-component free (ADCF), hydrolysate-free and regulatory-friendly cell culture medium. The versatility of this medium allows high transfection efficiency, consistent cell growth, high viable cell density, and high viral titers across many HEK 293 cell lines and AAV serotypes.
HyClone™ peak expression medium supports a seamless transition from growth to production of AAV using standard cell lines and processes. HyClone™ peak expression medium is available in liquid and powder formats in user-friendly packaging.
- Ready-to-use medium contains stable L-glutamine and poloxamer 188
- Designed for high cell yield and robust virus production
- Allows for direct adaptation
- Designed for small- to large-scale transfection and production applications
- Compatible with a variety of PEI- and lipid-based transfection reagents
Specifications
HEK | |
Viral Production | |
Mammalian | |
Cell Culture Media | |
0.2 μm Sterile-filtered | |
None |
Embryonic Kidney | |
Mammalian Cell Culture | |
Liquid | |
Sterile-filtered | |
Glutamine, Poloxamer 188 | |
1 L |