Learn More
Cytiva HyClone™ PF-CHO™ LS and PF-CHO MPS Media
Support growth of multiple CHO cell clones and production of a variety of recombinant proteins. HyClone™ PF CHO™ LS and PF CHO MPS Media is a protein-free media developed through Metabolic Pathway Design.
Brand: Cytiva SH30359.02
54.79 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 83
Additional Details : Weight : 1.36500kg
Description
Specifications
cGMP | |
PF CHO LS w/L-Glutamine (liquid) - Protein free | |
PF-CHO LS Liquid Medium | |
Glutamine, Poloxamer 188, Sodium Bicarbonate | |
1000 mL |
2° to 8°C, away from light | |
Liquid | |
0.2 μm Sterile-filtered | |
No Phenol Red, No HEPES |