Learn More
Cytiva HyClone™ PF-CHO™ LS and PF-CHO MPS Media
Support growth of multiple CHO cell clones and production of a variety of recombinant proteins. HyClone™ PF CHO™ LS and PF CHO MPS Media is a protein-free media developed through Metabolic Pathway Design.
Brand: Cytiva SH30333.02
190.95 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 83
Additional Details : Weight : 0.01250kg
Description
Specifications
cGMP | |
PF-CHO MPS (powder) - Protein free | |
Non-sterile | |
10 L |
2° to 8°C, away from light | |
Powder | |
No Glutamine, No Phenol Red, No Poloxamer 188, No HEPES, No Sodium Bicarbonate |