missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Porcine Serum, New Zealand Origin

Product Code. 10136173
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
This item is not returnable. View return policy

Product Code. 10136173

Brand: Cytiva SV30016.04

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Sourced from New Zealand and filtered in the United States through multiple, 200 nm pore size-rated filters. Serum is gamma irradiated at 25 to 40 kGy after filtration.

TRUSTED_SUSTAINABILITY

Specifications

Age >6 months
Country of Origin New Zealand
Form Liquid
Product Type Porcine Serum
Quantity 1000 mL
Sterilization Method Sequential Triple-filtered, 200 nm
Processing Irradiated (25 to 40 kGy)
Endotoxin Concentration ≤ 100 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 50 mg/dL
Product Line HyClone
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.