Learn More
GE Healthcare Hyclone™ HyClone™ SFM4CHO Media
Brand: GE Healthcare Hyclone™ SH30518.02
Code : A0
Additional Details : Weight : 10.00000kg
Specifications
cGMP | |
SFM4CHO Powder Media | |
1 x 10 L |
Powder | |
No Glutamine, No Phenol Red |