missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ SFM4HEK293™ Media
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
1000mL
500mL
Description
Support production of adenoviral vectors and recombinant proteins in HEK 239 cultures with HyClone™ SFM4HEK293™ Media, a versatile, protein-free cell culture media containing no animal-derived components.
Specifications
Specifications
| Form | Liquid |
| With Additives | Glutamine |
| Quantity | 500 mL |
| Certifications/Compliance | cGMP |
| Content And Storage | 2° to 8°C |
| Description | SFM4HEK293 Media |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction