Learn More
Cytiva HyClone™ SFM4Insect™ Media
Support growth of many key insect cell lines and production of a variety of recombinant proteins. HyClone™ SFM4Insect™ Media is free of animal-derived components (ADCF).
Brand: Cytiva SH30913.02
65.30 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : A0
Additional Details : Weight : 0.99000kg
Description
- Format: Liquid
Specifications
cGMP | |
SFM4Insect with L-Glutamine, Liquid | |
Insect Cell Medium | |
Glutamine, Poloxamer 188, Sodium Bicarbonate | |
1000 mL |
2° to 8°C, away from light | |
Liquid | |
0.2 μm Sterile-filtered | |
No Protein, No Phenol Red, No HEPES |