Learn More
Cytiva HyClone™ SFM4Insect Powdered Cell Culture Media
ADCF (animal-derived component-free) medium supports recombinant protein production with multiple insect cell lines.
Brand:  Cytiva SH30912.01
Product Code. 12954095
  • £242.00 / 5 L
Explore more special offers
Description
Description
SFM4Insect medium provides excellent growth of many key insect cell lines while requiring minimal adaptation. Additionally, SFM4Insect medium has been successfully tested in a variety of culture systems up to and including high density bioreactors.
  • For use with the baculovirus expression vector system (BEVS)
  • Designed for high cell yield and recombinant protein production
  • Complexed lipids for enhanced stability
  • Supports multiple insect cell lines
  • Supports direct or sequential adaptation
  • Manufactured using ISO 9001 and ISO 13485 certified processes according to 21 CFR 820 medical device current good manufacturing practices (cGMP)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 to 8°C, away from light and moisture
Non-sterile
No Sodium Bicarbonate
Powder
Glutamine
5 L