Learn More
Cytiva HyClone™ SFM4Transfx-293 Powdered Media
Promotes transfection using lipofection, polymer, or similar methods in HEK 293 cell culture.
Brand: Cytiva SH30861.01
Code : 83
Additional Details : CAS Number : 72-18-4 Weight : 0.10000kg
Description
HyClone SFMTransfx-293 medium is a serum-free and animal-derived component-free medium designed to support the growth of HEK 293 cell cultures and promote transfection using lipofection, polymer, or similar methods. This regulatory-friendly medium was developed to support high transfection efficiency, cell density, and productivity in suspension cultures.
SFM4Transfx-293 medium is available in liquid and powder formats.
- Supports high transfection efficiency.
- Allows for direct or sequential adaptation.
- Animal-derived component-free.
- Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities.
- Designed for high cell yield and recombinant protein/vector production.
Specifications
2 to 8°C, away from light and moisture | |
Non-sterile | |
5 L |
Powder | |
No Glutamine, No HEPES, No Phenol Red, No Sodium Bicarbonate |