Learn More
Cytiva HyClone™ SG 200 solution
Stable dipeptide of L-Alanyl L-Glutamine which acts as nutrient media supplement.
Brand: Cytiva SH30590.01
Code : 83
Additional Details : Weight : 0.31500kg
Description
- Used as nutrient media supplement
Specifications
cGMP and ISO 91:2 Guidelines | |
SG-200, (Stable dipeptide of L-AlanyL-Glutamine) 200mM, in 0.85% NaCl Solution | |
0.1 μm Sterile-filtered | |
100 mL |
2° to 8°C | |
2mM | |
=1.0EU/mL |