missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Tetracycline Screened Characterized Fetal Bovine Serum, US Origin
FBS with undetectable levels of tetracycline
£653.00 - £674.00
Specifications
Country of Origin | United States |
---|---|
Form | Liquid |
Specialty Type | Tetracycline screened |
Quantity | 500 mL |
Endotoxin Concentration | ≤ 10 EU/mL |
Product Code | Brand | Processing | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Processing | Price | Quantity & Availability | |||||
17692065
|
Cytiva
SH30071.03TH |
Heat Inactivated |
£653.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17602075
|
Cytiva
SH30071.03TIH25-40 |
Heat Inactivated, Gamma Irradiated (25 to 40 doses) |
£674.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tetracycline Screened FBS is designed for researchers using tetracycline-regulated gene expression systems in cultured cells and is suitable for expression studies in Tet-on/Tet-off systems.Specifications
United States | |
Tetracycline screened | |
≤ 10 EU/mL | |
≤ 10 mg/dL |
Liquid | |
500 mL | |
Yes |