missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Tetracycline Screened Fetal Bovine Serum
Click to view available options
Heat Inactivated:
No
Yes
Unit Size:
500mL
Specifications
Specifications
| Country of Origin | United States |
| Quantity | 500 mL |
| Sterility | Sterile |
| Volume (Metric) | 500 mL |
| Heat Inactivated | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction