missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Adult Bovine Serum, New Zealand Origin
Conformation Tested According to the European Agency for the Evaluation of Medicinal Products (EMEA), average age at time of collection is 6 to 8 years
£88.40 - £267.00
Specifications
Age | 6 to 8 years |
---|---|
Country of Origin | New Zealand |
Product Type | Adult Bovine Serum |
Sterilization Method | Triple-filtered |
Product Line | HyClone™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16635765
|
Cytiva
SH30528.03 |
1000 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16655765
|
Cytiva
SH30528.05 |
3000 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16625765
|
Cytiva
SH30528.02NI |
500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16645765
|
Cytiva
SH30528.03NI |
1000 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16665765
|
Cytiva
SH30528.05NI |
3000 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
6 to 8 years | |
Adult Bovine Serum | |
HyClone™ |
New Zealand | |
Triple-filtered | |
Tested According to European Pharmacopoeia Protocols |