missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Trypsin Protease

Derived from porcine pancreas. HyClone™ Trypsin Protease is used to disaggregate adherent cells from substrate or surface to which they are attached.

Brand:  Cytiva SH30042.01

11.11 GBP valid until 2025-12-31
Use promo code "24335" to get your promotional price.



 View more versions of this product

Product Code. 10366662

  • £22.60 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Derived from porcine pancreas
  • Gamma irradiated prior to hydration and filling
  • Formulated without calcium and magnesium
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store frozen
Liquid
Trypsin, porcine, Hanks’ Balanced Salt solution (HBSS), EDTA
2.5 g porcine trypsin (1:250/L gamma irradiated) in HBSS with 0.2 g/L EDTA
Porcine pancreas
0.25% (1 X)
0.1 μm Sterile-filtered
No Calcium, No Magnesium
100 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • TRYPSIN 0.25% EDTA

Signal Word
  • Danger

Hazard Category
  • Respiratory sensitiser Category 1

Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.

SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva HyClone™ Trypsin Protease > Trypsin 0.25%; 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.