missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GE Healthcare Hyclone™ HyClone™ US Donor Equine
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
Donor equine serum is carefully collected from selected donor herds that receive regular veterinary inspection and care. The animals are fed an enriched diet to ensure proper nutrition and are fasted prior to bleeding to reduce lipid concentration. The horses are bled in an enclosed facility that is specially equipped for aseptic collection. Coggins tests for equine infectious anaemia are routinely performed on each horse. Donor equine serum is sterile filtered using three sequential 100nm (0.1μm) pore size rated filters.
Specifications
Specifications
| Additional Assays | Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose |
| Species | Equine |
| Product Type | Equine Serum |
| Quantity | 500 mL |
| Sterility | Sterile |
| Sterilization Method | Sequential Triple-filtered, 100 nm |
| Tested For | BVDV, Rabies Virus, Reovirus, Cytopathogenic Agents |
| Heat Inactivated | Yes |
| Test Methods | Coggins |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu