missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Water for Injection (WFI) Quality Water

Product Code. 11531861
Click to view available options
Quantity:
500 mL
1 L
10 L
20 L
200 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Supports cell culture growth and biomanufacturing process.

  • Format: Liquid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
SMILES O

Specifications

IUPAC Name water
Molecular Weight (g/mol) 18.02
Quantity 10 L
Chemical Name or Material Water
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.