missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Water for Injection (WFI) Quality Water
Supports cell culture growth and biomanufacturing process.
£11.65 - £1980.00
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | dihydrogen oxide, dihydrogen monoxide |
SMILES | O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10464292
|
Cytiva
SH30221.17 |
500 mL | N/A | ||||||
10317002
|
Cytiva
SH30221.10 |
1 L | N/A | ||||||
11531861
|
Cytiva
SH30221.24 |
10 L |
£264.00
10 L |
In Stock
Log in to see stock. |
|||||
11541861
|
Cytiva
SH30221.25 |
20 L | N/A | ||||||
12329882
|
Cytiva
SH30221.28 |
200 L |
£1980.00
200 L |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
Description
- Format: Liquid
Chemical Identifiers
7732-18-5 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
O |
H2O | |
dihydrogen oxide, dihydrogen monoxide |
Specifications
7732-18-5 | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.02 |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Water |
Safety and Handling
missing translation for 'shelfLife' : 36 Months