missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 27-600 Nitrile Coated Heavy Duty Work Gloves - Knitwrist

Product Code. 12610792
Click to view available options
Size:
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves feature a unique coating formulation that provides increased resistance against sharp and abrasive environments, in addition to oil and grease.

  • Designed to be extra tough for use in environments that involve rough, abrasive materials
  • Constructed of cotton and jersey fabric for a high level of user comfort
  • Unique nitrile coating provides barrier protection against oil and grease
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Construction Cut & Sewn
Coating Material Nitrile
Color Blue, Natural
Cut Resistance Rating ANSI A1
Hand Specific
Material Cotton
Product Type Dip-coated Gloves
Size 10
EN388 Standard 4121B
For Use With (Application) Automotive Assembly, Machinery/Equipment Operation, and Metal Fabrication
Glove Cuff Style Knit
Series ActivArmr™ 27-600
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'