missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 27-602 Fully Nitrile Coated Heavy Duty Cotton Jersey Gloves

Product Code. p-8005277 Shop All Ansell Products
Click to view available options
Size:
8
9
10
Unit Size:
12 pairs

Product Code. 12660792

Brand: Ansell 27602080

Only null left
Add to basket
Add to basket

Ansell's unique nitrile coating formulation offers resistance to sharp/abrasive materials, oil, and grease. The heavy duty design is durable, and certified clean for metals and motor manufacturing.

  • Ansell's unique nitrile formulation offers resistance to sharp and abrasive materials
  • Gloves provides a barrier against grease and oil that will not degrade
  • Guaranteed clean for metals and motor manufacturing
  • Three times more durable than heavy-duty leather
  • Stronger and more flexible than PVC
  • Silicone-free
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Construction Cut and Sewn
Cuff Material Nitrile
Coating Material Nitrile
Color Blue, Natural
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Cotton
Product Type Dip-coated Gloves
Size 8
For Use With (Application) Automotive, Machinery/Equipment, and Metal Fabrication
Glove Cuff Style Knit
Series 27-602
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.