Learn More
Ansell ActivArmr™ 27-600 Nitrile Coated Heavy Duty Work Gloves - Safety Cuff
Gloves feature a unique coating formulation that provides increased resistance against sharp and abrasive environments, in addition to oil and grease.
£59.00
Specifications
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Construction Cut and Sewn
Coating Material Nitrile
Color Blue, Natural
View More Specs
Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12600802
View Documents
Ansell
27607080
8
£59.00
12 pairs
Estimated Shipment: 13-03-2025
to see stock.
Add to basket
12610802
View Documents
Ansell
27607090
9
£59.00
12 pairs
In Stock
to see stock.
Edge
Add to basket
12680792
View Documents
Ansell
27607100
10
£59.00
12 pairs
Estimated Shipment: 13-03-2025
to see stock.
Add to basket
12690792
View Documents
Ansell
27607110
11
£59.00
12 pairs
Estimated Shipment: 13-03-2025
to see stock.
Add to basket
Description
Description
  • Designed to be extra tough for use in environments that involve rough, abrasive materials
  • Constructed of cotton and jersey fabric for a high level of user comfort
  • Unique nitrile coating provides barrier protection against oil and grease
  • Safety cuff increases protection to the wrist
Specifications
Specifications
ANSI 4
Cut and Sewn
Blue, Natural
Specific
Dip-coated Gloves
Automotive Assembly, Machinery/Equipment Operation, and Metal Fabrication
ActivArmr™ 27-607
EN 420:2003 + A1:2009; Category II
Nitrile
ANSI A1
Cotton
4121B
Safety
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Ansell ActivArmr™ 27-600 Nitrile Coated Heavy Duty Work Gloves - Safety Cuff