missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydantoin, 99%, Thermo Scientific Chemicals
£25.30 - £113.00
Chemical Identifiers
CAS | 461-72-3 |
---|---|
Molecular Formula | C3H4N2O2 |
Molecular Weight (g/mol) | 100.08 |
MDL Number | MFCD00005259 |
InChI Key | WJRBRSLFGCUECM-UHFFFAOYSA-N |
Synonym | hydantoin, 2,4-imidazolidinedione, glycolylurea, imidazole-2,4 3h,5h-dione, dantochlor, 2,4 3h,5h-imidazoledione, hydantoine, ccris 6532, 2-imidazolin-4 or 5-one, 2-hydroxy, 4h-imidazole-2,5-diol |
PubChem CID | 10006 |
ChEBI | CHEBI:27612 |
IUPAC Name | imidazolidine-2,4-dione |
SMILES | O=C1CNC(=O)N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10469090
|
Thermo Scientific Acros
168701000 |
100 g | Plastic bottle |
£25.30
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10134280
|
Thermo Scientific Acros
168705000 |
500 g | Plastic bottle |
£113.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
461-72-3 | |
100.08 | |
WJRBRSLFGCUECM-UHFFFAOYSA-N | |
10006 | |
imidazolidine-2,4-dione |
C3H4N2O2 | |
MFCD00005259 | |
hydantoin, 2,4-imidazolidinedione, glycolylurea, imidazole-2,4 3h,5h-dione, dantochlor, 2,4 3h,5h-imidazoledione, hydantoine, ccris 6532, 2-imidazolin-4 or 5-one, 2-hydroxy, 4h-imidazole-2,5-diol | |
CHEBI:27612 | |
O=C1CNC(=O)N1 |
Specifications
461-72-3 | |
White to Yellow | |
99% | |
MFCD00005259 | |
15, 4799 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alkali hydroxide solutions,soluble in alkali,slightly soluble in ether | |
O=C1CNC(=O)N1 | |
100.08 | |
CHEBI:27612 | |
99% | |
Hydantoin, 99% |
220°C to 223°C | |
Authentic | |
C3H4N2O2 | |
24, 242 | |
hydantoin, 2,4-imidazolidinedione, glycolylurea, imidazole-2,4 3h,5h-dione, dantochlor, 2,4 3h,5h-imidazoledione, hydantoine, ccris 6532, 2-imidazolin-4 or 5-one, 2-hydroxy, 4h-imidazole-2,5-diol | |
WJRBRSLFGCUECM-UHFFFAOYSA-N | |
imidazolidine-2,4-dione | |
10006 | |
100.08 | |
Fine Powder |
Safety and Handling
missing translation for 'einecsNumber' : 207-313-3
missing translation for 'rtecsNumber' : MT8210000
missing translation for 'tsca' : TSCA
RUO – Research Use Only