missing translation for 'onlineSavingsMsg'
Learn More

HYDRA-POINT™ Coulometric Vessel Solution, BAKER ANALYZED™ Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

£341.00 - £653.00

Specifications

Flash Point 11.00°C
Chemical Name or Material HYDRA-POINT™ Coulometric Vessel Solution
Packaging Narrow Mouth Amber Glass Bottle
Grade BAKER ANALYZED™ Reagent
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15517634
View Documents
Avantor J.T.Baker
6280-01
500 mL Narrow Mouth Amber Glass Bottle
£341.00
500mL
Please to purchase this item. Need a web account? Register with us today!
15527634
View Documents
Avantor J.T.Baker
6280-02
1 L Narrow Mouth Amber Glass Bottle
£653.00
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

    For Karl Fischer titration
Specifications

Specifications

11.00°C
HYDRA-POINT™ Coulometric Vessel Solution
Narrow Mouth Amber Glass Bottle
BAKER ANALYZED™ Reagent
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H351-Suspected of causing cancer.
  • H370-Causes damage to organs.
  • H373-May cause damage to organs.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.

SDS
Documents

Documents

Product Certifications