missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRA-POINT™ Coulometric Vessel Solution, CFC Free, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
£484.00 - £1530.00
Specifications
Flash Point | 55.60°C |
---|---|
Chemical Name or Material | HYDRA-POINT™ Coulometric Vessel Solution |
Name Note | CFC Free |
Packaging | Narrow Mouth Amber Glass Bottle |
Grade | BAKER ANALYZED™ Reagent |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15577634
|
Avantor J.T.Baker
6284-01 |
500 mL | Narrow Mouth Amber Glass Bottle |
£484.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15587634
|
Avantor J.T.Baker
6284-02 |
1 L | Narrow Mouth Amber Glass Bottle |
£1530.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For Coulometric Karl Fischer Analysis
Specifications
55.60°C | |
CFC Free | |
BAKER ANALYZED™ Reagent |
HYDRA-POINT™ Coulometric Vessel Solution | |
Narrow Mouth Amber Glass Bottle |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H361-Suspected of damaging fertility or the unborn child.
- H370-Causes damage to organs.
- H373-May cause damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HYDRA-POINT™ Coulometric Vessel Solution, CFC Free, BAKER ANALYZED™ Reagent, J.T. Baker™