missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™-Buffer for Acids, Liquid buffer medium for Karl Fischer titration in acidic samples (based on imidazole), Fluka, Honeywell
Click to view available options
Quantity:
6 x 500 mL
Description
Advantages of HYDRANAL- Auxiliaries
- Broad product range of solubilizer for special sample application
- Broad product range of buffer for pH related sample application
- Drying agents for cell protection and drying of carrier gasses.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 0.960 g/cm3 (20°C) |
Flash Point | 11°C |
Quantity | 6 x 500 mL |
CAS | 7446-09-5, 288-32-4, 67-56-1 |
CAS Min % | ≥5.00, ≥30.00, ≥50.00 |
CAS Max % | <10.00, <50.00, <70.00 |
Boiling Point | 65°C (1.013 hPa).The physical data is that of the main component. |
Packaging | Polycoated Bottle |
Physical Form | Liquid |
Molecular Formula | O2S |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- HYDRANAL Buffer for Acids
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Reproductive toxicity Category 1B
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H314-Causes severe skin burns and eye damage.
- H360D-May damage the unborn child.
- H370-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : methanol, imidazole,sulphur dioxide
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'