missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-Buffer Base, Honeywell Fluka™

Product Code. 15615170 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
6 x 1 L
Unit Size:
1 L
Pack of 6
This item is not returnable. View return policy

Product Code. 15615170

Brand: Honeywell-Hydranal 378596X1L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Liquid buffer medium for Karl Fischer titration in alkaline samples (based on salicylic acid)

Advantages of HYDRANAL- Auxiliaries

  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Buffer Base
CAS 69-72-7, 67-56-1, 64-17-5, 121-44-8
Quantity 6 x 1 L
Flash Point 11°C (51.8°F)
MDL Number MFCD00147178
UN Number UN1993BE
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL Buffer for Bases
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H361d-Suspected of damaging the unborn child.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : ethanol, methanol, salicylic acid, triethylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.