missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Composite 2, Reagent for volumetric one-component Karl Fischer titration (methanol free), Honeywell™ Fluka™

Product Code. p-7072135 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 500 mL
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
500mL
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15624450

Brand: Honeywell-Hydranal 34806500ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Chemical Name or Material HYDRANAL™-Composite 2
CAS 7446-09-5, 693-98-1, 288-32-4, 111-90-0, 68007-08-9
Density 1.11g/mL
Quantity 500 mL
Flash Point 90°C (194°F)
compliance-icons
Product Identifier
  • HYDRANAL-Composite 2
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : imidazole, sulphur dioxide, iodine, 2-Methylimidazole, 1H-Imidazole monohydriodide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.