missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Composite 5, Honeywell Fluka™
Titrating agent for volumetric, one-component Karl Fischer titrations (Methanol-free)
£138.19 - £751.72
Specifications
Flash Point | 99°C |
---|---|
Chemical Name or Material | HYDRANAL™ - Composite 5 |
Color | Brown |
CAS | 111-90-0 |
pH | 4.5 to 5.5 (at 20°C) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16841910
|
Honeywell Fluka
34805-1L-OM |
1 L |
£138.19
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16895211
|
Honeywell Fluka
34805-6X1L-OM |
6 x 1 L |
£751.72
6 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Improved Composition
- Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE)
- Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator’s performance
Advantages of Hydranal One-Component Reagents:
- Unlimited water capacity
- Convenient and easy to use
- The greatest flexibility in working media selection
- Reduced toxicity compared to methanol based systems
- Suitable for methanol-reacting compounds (ex. ketones and aldehydes)
- 3 year shelf life
Specifications
99°C | |
Brown | |
4.5 to 5.5 (at 20°C) | |
Metrohm OMNIS Smart Reagent | |
DE Solvent; 2-(2-Ethoxyethoxy)ethanol; CARBITOL; Diethylene glycol ethyl ether; Ethyldiglycol | |
Volumetric One-Component KF Titration |
HYDRANAL™ - Composite 5 | |
111-90-0 | |
194°C (at 1.013 hPa) | |
Liquid | |
Completely Miscible in Water |
Safety and Handling
Recommended Storage : Ambient
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HYDRANAL NEXTGEN Composite 5 OM
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H351-Suspected of causing cancer.
- H360D-May damage the unborn child.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 2-(2-ethoxyethoxy)ethanol, sulphur dioxide, 1H-imidazole monohydroiodide, iodine, imidazole, 2-methy
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title