Learn More
HYDRANAL™ - Composite 5, Reagent for volumetric one-component Karl Fischer titration (methanol free), Honeywell™ Fluka™
Brand: Honeywell Fluka 34805-500ML
76.52 GBP valid until 2024-12-31
Use promo code "21961" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.58500kg
Specifications
90°C (194°F) | |
HYDRANAL™-Composite 5 | |
111-90-0,7553-56-2,288-32-4,7446-09-5,68007-08-9,693-98-1 |
500 mL | |
1.15g/mL |
Safety and Handling
- HYDRANAL-Composite 5
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- Carcinogenicity Category 2
Hazard Statement
- H351-Suspected of causing cancer.
- H360D-May damage the unborn child.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : sulphur dioxide, 1H-Imidazole monohydriodide, iodine, imidazole, 2-Methylimidazole