Learn More
HYDRANAL™ - Composite 5 K, Reagent for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free), Honeywell™ Fluka™
Brand: Honeywell-Hydranal 34816-500ML
70.65 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description

Specifications
HYDRANAL™-Composite 5 K | |
90°C (194°F) | |
111-90-0, 7553-56-2, 288-32-4, 693-98-1, 7446-09-5, 68007-08-9 |
1.15g/mL | |
500 mL |
Safety and Handling
- HYDRANAL
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- Carcinogenicity Category 2
Hazard Statement
- H351-Suspected of causing cancer.
- H360D-May damage the unborn child.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 2-(2-Ethoxyethoxy)ethanol, iodine, imidazole, 2-Methylimidazole, sulphur dioxide, 1H-Imidazole monoh
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Your input is important to us. Please complete this form to provide feedback related to the content on this product.