missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-CompoSolver E, Honeywell Fluka™

Medium for volumetric one-component Karl Fischer titration (ethanol-based)

Brand:  Honeywell Fluka 34734-6X1L

Code : NEW

Additional Details : CAS Number : 693-98-1 Weight : 4.81200kg Transport : UN number : 1170 Chem class : 3 Pack group : II


 View more versions of this product

Product Code. 15654380

  • £305.68 / Pack of 6
In Stock
to see stock.

Description

Description

Advantages of Hydranal one-component reagents:

  • Unlimited water capacity
  • Convenient and easy to use
  • The greatest flexibility in working media selection
  • reduced toxicity compared to methanol based systems
  • Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
  • Long shelf life (three years)

Improved Composition

Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator’s performance.

Specifications

Specifications

12°C (53.6°F)
HYDRANAL™-CompoSolver E
7446-09-5,693-98-1,64-17-5
6 x 1 L
0.80g/mL
UN1170
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : ethanol; ethyl alcohol, 2-Methylimidazole, sulphur dioxide

SDS Documents

Documents

Certificates
Special Offers

Special Offers