Learn More
HYDRANAL™ - Coulomat AF 7, For coulometer AF7, Honeywell Fluka™
for coulometer AF7
Brand:  Honeywell-Hydranal 34829-1L
 View more versions of this product
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15674490
  • £202.11 / 1 L
Explore more special offers
Description
Description
Advantages of Hydranal Coulometric Reagents
  • Easy to use
  • High accuracy for trace amounts of water
  • Stable conditions of the titration vessel
  • Broad product range
  • Long shelf life (up to five years)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HYDRANAL™-Coulomat AF 7
1 L
Glass bottle
11°C (51.8°F)
67-56-1, 7446-09-5, 288-32-4
UN1230