missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Coulomat AG-Oven, Reagent for coulometric KF titration with oven (anolyte solution), for cells with and without diaphragm, Honeywell Fluka

Brand:  Honeywell-Hydranal 34739-500ML

105.51 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.



 View more versions of this product

Product Code. 15614390

  • £111.06 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HYDRANAL™ - Coulomat AG-Oven
0.980 g/cm3
500 mL
<70.0000%, <30.0000%, <20.0000%, <10.0000%, <10.0000%, <10.0000%
64°C
UN1230
Reagent for coulometric KF titration with oven (anolyte solution), for cells with and without diaphragm
13°C
≥50.0000%, ≥20.0000%, ≥10.0000%, ≥5.0000%, ≥5.0000%, ≥5.0000%
57-55-6, 111-42-2, 288-32-4, 7446-09-5, 68007-08-9
Glass Bottle
Karl Fischer
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

    Days

    0

    Hours

    0

    Minutes

    25

    Seconds

    5
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

    Days

    30

    Hours

    0

    Minutes

    25

    Seconds

    5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
HYDRANAL™ - Coulomat AG-Oven, Reagent for coulometric KF titration with oven (anolyte solution), for cells with and without diaphragm, Honeywell Fluka >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.