missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Coulomat AK, Reagent for coulometric KF titration in ketones (anolyte solution), preferred for cells with diaphragm, Honeywell Fluka
£192.22 - £1010.04
Specifications
Density | 1.200 g/cm3 |
---|---|
Flash Point | 32°C |
Chemical Name or Material | HYDRANAL™ - Coulomat AK |
Name Note | Reagent for coulometric KF titration in ketones (anolyte solution), preferred for cells with diaphragm |
CAS | 109-86-4, 67-66-3, 75-89-8, 101023-55-6, 68007-08-9, 7446-09-5 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15684480
|
Honeywell-Hydranal
34820-500ML |
500 mL | Glass Bottle |
£192.22
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15694480
|
Honeywell-Hydranal
34820-6X500ML |
6 x 500 mL | Glass Bottle |
£1010.04
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Specifications
1.200 g/cm3 | |
HYDRANAL™ - Coulomat AK | |
109-86-4, 67-66-3, 75-89-8, 101023-55-6, 68007-08-9, 7446-09-5 | |
<50.0000%, <30.0000%, <20.0000%, <10.0000%, <5.0000%, <5.0000% | |
Glass Bottle | |
Karl Fischer |
32°C | |
Reagent for coulometric KF titration in ketones (anolyte solution), preferred for cells with diaphragm | |
≥30.0000%, ≥20.0000%, ≥10.0000%, ≥5.0000%, ≥1.0000%, ≥1.0000% | |
60°C | |
UN1992 |
Safety and Handling
P201-P210-P280-P304 + P340 + P310-P305 + P351 + P338-P308 + P311
missing translation for 'shelfLife' : 1800 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Flammable liquids, toxic, n.o.s.
Recommended Storage : Room Temp
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HYDRANAL
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H360FD-May damage fertility. May damage the unborn child.
- H372-Causes damage to organs.
- H351-Suspected of causing cancer.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 2-methoxyethanol; ethylene glycol monomethyl ether, trichloromethane; chloroform, 2,2,2-Trifluoroeth
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title