missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Coulomat CG, Reagent for coulometric KF titration (catholyte solution), Honeywell Fluka™

Brand:  Honeywell-Hydranal 34840-6X50ML

426.51 GBP valid until 2024-11-15
Use promo code "21643" to get your promotional price.



 View more versions of this product

Product Code. 15674500

  • £480.19 / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HYDRANAL™ - Coulomat CG
1.064 g/cm3
1 Kit
≥50.0000%, ≥30.0000%, ≥1.0000%, ≥1.0000%
63°C
UN1230
Reagent for coulometric KF titration (catholyte solution)
14°C
14426-21-2, 67-56-1, 134227-25-1, 7446-09-5
<70.0000%, <50.0000%, ≤5.0000%, <5.0000%
Ampoule
Karl Fischer
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HYDRANAL-Coulomat CG

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Diethanolamine hydrochloride, methanol,Ethanol, 2,2'-iminobis-, hydriodide, sulphur dioxide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers