missing translation for 'onlineSavingsMsg'
Learn More
HYDRANAL™ - Formamide dry, Solvent for KF titration, Honeywell Fluka™ Product Code.: 15644360

HYDRANAL™ - Formamide dry, Solvent for KF titration, Honeywell Fluka™

Product Code. 15644360 Shop All Honeywell Fluka Products
1 L
Click to view available options
Quantity:
1 L
6 x 1 L
This item is not returnable. View return policy

Product Code. 15644360

Brand: Honeywell-Hydranal 347241L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Solvent for Karl Fischer titration

Advantages of HYDRANAL- Auxiliaries

  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

Density 1.134g/mL at 25°C
Flash Point 175°C (347°F)
Quantity 1 L
Vapor Density 1.55 (vs air)
Linear Formula HCONH2
UN Number NONH for all modes of transport
Beilstein 505995
Formula Weight 45.04g/mol
Chemical Name or Material HYDRANAL™-Formamide dry
compliance-icons
Product Identifier
  • HYDRANAL-Formamide dry
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : formamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
HYDRANAL™ - Formamide dry, Solvent for KF titration, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.