missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - KetoSolver, Reagent for volumetric one-component KF titration in aldehydes and ketones (working medium), Honeywell Fluka™

Medium for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free)

Brand:  Honeywell Fluka 34738-500ML

Code : NEW

Additional Details : CAS Number : 64-17-5 Weight : 0.45500kg Transport : UN number : 1993 Chem class : 3 Pack group : II


 View more versions of this product

Product Code. 15684380

  • £54.03 / 500mL
In Stock
to see stock.

Description

Description

Advantages of Hydranal one-component reagents:

  • Unlimited water capacity
  • Convenient and easy to use
  • The greatest flexibility in working media selection
  • reduced toxicity compared to methanol based systems
  • Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
  • Long shelf life (three years)

Improved Composition

Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator’s performance.

Specifications

Specifications

21°C (69.8°F)
HYDRANAL™-KetoSolver
107-98-2,64-17-5,288-32-4,101023-55-6
500 mL
0.91g/mL
UN1993BE
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HYDRANAL-KetoSolver

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H360D-May damage the unborn child.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : 1-methoxy-2-propanol; monopropylene glycol methyl ether, ethanol; ethyl alcohol,imidazole, Imidazol

SDS Documents

Documents

Certificates
Special Offers

Special Offers